Learn More
Cayman Chemical DIphthRIa ToxIn CRM197 mutn
SDP
Catalog No. 502993836
Encompass_Preferred
An inactive diphtheria toxin; contains a single base change in the structural gene resulting in the substitution of glutamic acid for glycine which blocks ADP-ribosylation; has no enzymatic activity but is immunologically indistinguishable from diphtheria toxin
Catalog No. 50-299-3836 Supplier Cayman Chemical Supplier No. 23218250 UG
An inactive diphtheria toxin; contains a single base change in the structural gene resulting in the substitution of glutamic acid for glycine which blocks ADP-ribosylation; has no enzymatic activity but is immunologically indistinguishable from diphtheria toxin