Learn More
Cayman Chemical DIphthRIa ToxoId from Coryn
SDP
Catalog No. 502964261
Encompass_Preferred
An inactive diphtheria toxin; non-specifically cross linked and subject to rearrangement
Catalog No. 50-296-4261 Supplier Cayman Chemical Supplier No. 23219500 UG
An inactive diphtheria toxin; non-specifically cross linked and subject to rearrangement