Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(+)-Dipivaloyl-D-tartaric Acid 98.0+%, TCI America™
SDP

Catalog No. D27025G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D27025G 5 g
D27021G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D27025G Supplier TCI America Supplier No. D27025G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 76769-55-6 | C14H22O8 | 318.32 g/mol

Chemical Identifiers

CAS 76769-55-6
Molecular Formula C14H22O8
Molecular Weight (g/mol) 318.32
MDL Number MFCD00015634
InChI Key UFHJEZDFEHUYCR-YZYOREDDNA-N
PubChem CID 16212279
IUPAC Name (2S,3S)-2,3-bis[(2,2-dimethylpropanoyl)oxy]butanedioic acid
SMILES CC(C)(C)C(=O)O[C@@H]([C@H](OC(=O)C(C)(C)C)C(O)=O)C(O)=O

Specifications

Melting Point 150°C
Color White
Quantity 5 g
Synonym +-dipivaloyl-d-tartaric acid, 2s,3s-2,3-bis pivaloyloxy succinic acid, d-+-dipivaloyl tartaric acid, pubchem8173, --bis 2,2-dimethylpropionyl-l-tartaric acid, dipivaloyl-l-tartaric acid, +-dipivaloyl-d-tartaricacid, +-dpta, ?-o,o'-di-pivaloyl-l-tartaric acid, +-o,o'-di-pivaloyl-d-tartaric acid
Formula Weight 318.32
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material (+)-Dipivaloyl-D-tartaric Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.