Learn More
Medchemexpress LLC Dipivaloylmethane | 1118-71-4 | 184.28 | 25 G
SDP
Catalog No. 5000402808
Encompass_Preferred
Dipivaloylmethane is a versatile biochemical reagent for life science research, functioning as both a biological material and an organic compound.
  • Used in the synthesis of α-aryl-β-diketones
  • Used in the synthesis of dicyanamidobenzene-bridge diruthenium complex
Catalog No. 50-004-02808 Supplier Medchemexpress LLC Supplier No. 25GHY34417
Dipivaloylmethane is a versatile biochemical reagent for life science research, functioning as both a biological material and an organic compound.
  • Used in the synthesis of α-aryl-β-diketones
  • Used in the synthesis of dicyanamidobenzene-bridge diruthenium complex