Learn More
Medchemexpress LLC Dipivaloylmethane | 1118-71-4 | 98.74% | 184.28 | 50 G
SDP
Catalog No. 5000404056
Encompass_Preferred
Dipivaloylmethane is a biochemical reagent that can be used as a biological material or organic compound for life science related research. It is for research use only and not sold to patients.
  • Biochemical reagent
  • Can be used as a biological material or organic compound for life science related research
  • Used in the synthesis of α-aryl-β-diketones
  • Used in the synthesis of dicyanamidobenzene-bridge diruthenium complex
Catalog No. 50-004-04056 Supplier Medchemexpress LLC Supplier No. HY3441750G
Add to Cart
Add to Cart
Dipivaloylmethane is a biochemical reagent that can be used as a biological material or organic compound for life science related research. It is for research use only and not sold to patients.
  • Biochemical reagent
  • Can be used as a biological material or organic compound for life science related research
  • Used in the synthesis of α-aryl-β-diketones
  • Used in the synthesis of dicyanamidobenzene-bridge diruthenium complex