Learn More
Medchemexpress LLC Dipivefrin-d6 hydrochloride | 64019-93-8 | 99.7% | 1 MG
SDP
Catalog No. 5000411975
Encompass_Preferred
Dipivefrin-d6 hydrochloride is a deuterium-labeled version of Dipivefrin hydrochloride. It acts as an antiglaucoma proagent, converting into epinephrine in the cornea. This compound is valuable for quantitative analysis and drug development studies, where deuteration can influence pharmacokinetic and metabolic profiles.
  • Used as a tracer.
  • Functions as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Incorporates stable heavy isotopes for quantitation in drug development.
  • Deuteration can influence pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-004-11975 Supplier Medchemexpress LLC Supplier No. HYB1323S1MG
Dipivefrin-d6 hydrochloride is a deuterium-labeled version of Dipivefrin hydrochloride. It acts as an antiglaucoma proagent, converting into epinephrine in the cornea. This compound is valuable for quantitative analysis and drug development studies, where deuteration can influence pharmacokinetic and metabolic profiles.
  • Used as a tracer.
  • Functions as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Incorporates stable heavy isotopes for quantitation in drug development.
  • Deuteration can influence pharmacokinetic and metabolic profiles of drugs.