Learn More
Medchemexpress LLC Dipivefrin hydrochloride | 64019-93-8 | 98.4% | 100 MG
SDP
Catalog No. 5000412077
Encompass_Preferred
Dipivefrin hydrochloride is an antiglaucoma proagent that is hydrolyzed by esterases in the cornea to its active compound, epinephrine. It is for research use only and not sold to patients.
  • Antiglaucoma proagent
  • Hydrolyzed to active epinephrine by corneal esterases
  • Induces enhancement of memory involving central beta-adrenergic mechanisms in male 60-day-old CFW mice (0.3-10 μg/kg; i.p.), significantly enhancing retention of inhibitory avoidance and Y-maze discrimination tasks in a dose-dependent manner.
Catalog No. 50-004-12077 Supplier Medchemexpress LLC Supplier No. HYB1323100MG
Dipivefrin hydrochloride is an antiglaucoma proagent that is hydrolyzed by esterases in the cornea to its active compound, epinephrine. It is for research use only and not sold to patients.
  • Antiglaucoma proagent
  • Hydrolyzed to active epinephrine by corneal esterases
  • Induces enhancement of memory involving central beta-adrenergic mechanisms in male 60-day-old CFW mice (0.3-10 μg/kg; i.p.), significantly enhancing retention of inhibitory avoidance and Y-maze discrimination tasks in a dose-dependent manner.