Learn More
Medchemexpress LLC Dipivefrin hydrochloride | 64019-93-8 | MFCD00793796 | 98.4% | 1 ML
SDP
Catalog No. 5000412484
Encompass_Preferred
Dipivefrin hydrochloride is an antiglaucoma proagent that is hydrolyzed to epinephrine, the active compound, by esterases in the cornea. This product is for research use only and not intended for patient use.
  • Functions as an antiglaucoma proagent
  • Hydrolyzed to active compound epinephrine by corneal esterases
  • Intended for research use only
  • Store solid at -20°C under nitrogen, away from moisture
  • Store in solvent at -80°C for 6 months or -20°C for 1 month under nitrogen, away from moisture
Catalog No. 50-004-12484 Supplier Medchemexpress LLC Supplier No. HYB132310MM/1ML
Dipivefrin hydrochloride is an antiglaucoma proagent that is hydrolyzed to epinephrine, the active compound, by esterases in the cornea. This product is for research use only and not intended for patient use.
  • Functions as an antiglaucoma proagent
  • Hydrolyzed to active compound epinephrine by corneal esterases
  • Intended for research use only
  • Store solid at -20°C under nitrogen, away from moisture
  • Store in solvent at -80°C for 6 months or -20°C for 1 month under nitrogen, away from moisture