Learn More
Cayman Chemical DIpIvefrInhydrochlorIde 100mg
SDP
Catalog No. 502932564
Encompass_Preferred
A prodrug of epinephrine; reduces bovine primary trabecular meshwork cell density in vitro (IC50 = 115 µM); induces a fibroblast-like morphology and actin cytoskeleton disruption in cultured bovine primary trabecular meshwork cells (103 µM); enhances calcium signaling and induces cytotoxicity in cultured bovine corneal endothelial cells (28 µM); suppresses primary human corneal keratinocyte proliferation in vitro (280 µM)
Catalog No. 50-293-2564 Supplier Cayman Chemical Supplier No. 24028100 MG
A prodrug of epinephrine; reduces bovine primary trabecular meshwork cell density in vitro (IC50 = 115 µM); induces a fibroblast-like morphology and actin cytoskeleton disruption in cultured bovine primary trabecular meshwork cells (103 µM); enhances calcium signaling and induces cytotoxicity in cultured bovine corneal endothelial cells (28 µM); suppresses primary human corneal keratinocyte proliferation in vitro (280 µM)