Learn More
Cayman Chemical Dipivefrin (hydrochloride), Purity: ≥98%, CAS: 64019-93-8, 50 mg
SDP
Catalog No. 502935557
Encompass_Preferred
A prodrug of epinephrine; reduces bovine primary trabecular meshwork cell density in vitro (IC50 = 115 µM); induces a fibroblast-like morphology and actin cytoskeleton disruption in cultured bovine primary trabecular meshwork cells (103 µM); enhances calcium signaling and induces cytotoxicity in cultured bovine corneal endothelial cells (28 µM); suppresses primary human corneal keratinocyte proliferation in vitro (280 µM)
Catalog No. 50-293-5557 Supplier Cayman Chemical Supplier No. 2402850 MG
A prodrug of epinephrine; reduces bovine primary trabecular meshwork cell density in vitro (IC50 = 115 µM); induces a fibroblast-like morphology and actin cytoskeleton disruption in cultured bovine primary trabecular meshwork cells (103 µM); enhances calcium signaling and induces cytotoxicity in cultured bovine corneal endothelial cells (28 µM); suppresses primary human corneal keratinocyte proliferation in vitro (280 µM)