Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dipotassium Dihydrogen Ethylenediaminetetraacetic Acid, 99%, Spectrum™ Chemical
SDP

Catalog No. 18610282 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
500 g
2.5 kg
12 kg
50 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-610-282 500 g Poly Bottle
18-610-283 2.5 kg Poly Pail
18-610-284 12 kg Poly Pail
18-610-285 50 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18-610-282 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. E1042500GM
Add to Cart
Add to Cart

Chemical Identifiers

CAS 25102-12-9

Specifications

CAS Max % 1
pH 4.2 to 4.8
Loss on Drying 0.1
Packaging Poly Bottle
Quantity 500 g
Percent Purity 0.99
Grade Reagent

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.