Learn More
TARGETMOL CHEMICALS INC Dipotassium glycyrrhizinate
SDP
Catalog No. 502876996
Encompass_Preferred
Also available in 1 g, 1 mL, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Dipotassium glycyrrhizinate is an anionic surfactant having anti-allergic and anti-inflammatory activities. Purity 89.63%
Catalog No. 50-287-6996 Supplier TARGETMOL CHEMICALS INC Supplier No. T5666500MG
Also available in 1 g, 1 mL, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Dipotassium glycyrrhizinate is an anionic surfactant having anti-allergic and anti-inflammatory activities. Purity 89.63%