Learn More
Medchemexpress LLC Dipotassium hydrogen phosphate | 7758-11-4 | MFCD00011383 | 100.0% | 174.17 g·mol⁻¹ | HK2O4P | 50 G
SDP
Catalog No. 5000301703
Encompass_Preferred
Dipotassium hydrogen phosphate is an inorganic, dibasic potassium phosphate salt commonly used as a buffering agent and phosphate source in laboratory, biochemical, and formulation applications. It is supplied as a high-purity, water-soluble solid suitable for buffer preparation, pH control, and pharmaceutical excipient research.
  • High purity (99.95%) for analytical and research use.
  • Reliable buffering capacity for precise pH control.
  • Water soluble for easy buffer preparation.
  • Suitable as a phosphate source in biochemical and molecular biology workflows.
  • Stable solid form with extended shelf life when stored properly.
  • Available in multiple pack sizes for lab and scale-up needs.
Catalog No. 50-003-01703 Supplier Medchemexpress LLC Supplier No. HYW01988350G
Dipotassium hydrogen phosphate is an inorganic, dibasic potassium phosphate salt commonly used as a buffering agent and phosphate source in laboratory, biochemical, and formulation applications. It is supplied as a high-purity, water-soluble solid suitable for buffer preparation, pH control, and pharmaceutical excipient research.
  • High purity (99.95%) for analytical and research use.
  • Reliable buffering capacity for precise pH control.
  • Water soluble for easy buffer preparation.
  • Suitable as a phosphate source in biochemical and molecular biology workflows.
  • Stable solid form with extended shelf life when stored properly.
  • Available in multiple pack sizes for lab and scale-up needs.