Learn More
Medchemexpress LLC Dipotassium hydrogen phosphate | 7758-11-4 | MFCD00011383 | 100.0% | 174.176 g/mol | K2HPO4 | 100 G
SDP
Catalog No. 5000299971
Encompass_Preferred
Dipotassium hydrogen phosphate is an anhydrous, white crystalline salt used as a buffering agent and pH adjuster in laboratory and pharmaceutical formulations. It provides a dibasic phosphate source for preparing aqueous buffers, controlling ionic strength, and stabilizing pH in biological and chemical systems.
  • High purity (99.95%) suitable for research and formulation.
  • Functions as a buffering agent and pH adjuster in aqueous systems.
  • Soluble in water, enabling easy buffer preparation.
  • Anhydrous white crystalline form with stable storage properties.
  • Suitable as a pharmaceutical excipient and general laboratory reagent.
  • Compatible with common biological and chemical assays.
Catalog No. 50-002-99971 Supplier Medchemexpress LLC Supplier No. HYW019883100G
Dipotassium hydrogen phosphate is an anhydrous, white crystalline salt used as a buffering agent and pH adjuster in laboratory and pharmaceutical formulations. It provides a dibasic phosphate source for preparing aqueous buffers, controlling ionic strength, and stabilizing pH in biological and chemical systems.
  • High purity (99.95%) suitable for research and formulation.
  • Functions as a buffering agent and pH adjuster in aqueous systems.
  • Soluble in water, enabling easy buffer preparation.
  • Anhydrous white crystalline form with stable storage properties.
  • Suitable as a pharmaceutical excipient and general laboratory reagent.
  • Compatible with common biological and chemical assays.