Learn More
Medchemexpress LLC Dipotassium hydrogen phosphate | 7758-11-4 | MFCD00011383 | 100.0% | 174.176 g/mol | K2HPO4 | 500 G
SDP
Catalog No. 5000301507
Encompass_Preferred
Dipotassium hydrogen phosphate (K2HPO4, CAS 7758-11-4) is an anhydrous dibasic potassium phosphate salt used as a buffering agent, pH adjuster, and phosphate source in laboratory and pharmaceutical applications. It typically appears as a white crystalline powder with high purity and solubility in water, making it suitable for biochemical assays, media preparation, and formulation work requiring stable alkaline buffer capacity.
  • High purity (99.95%) suitable for analytical and formulation use.
  • Functions as a buffering agent and pH adjuster in aqueous systems.
  • Provides a soluble source of phosphate for media and reagent preparation.
  • Available in lab-scale pack sizes up to 500 g for routine use.
  • Stable, white crystalline powder with good water solubility.
Catalog No. 50-003-01507 Supplier Medchemexpress LLC Supplier No. HYW019883500G
Dipotassium hydrogen phosphate (K2HPO4, CAS 7758-11-4) is an anhydrous dibasic potassium phosphate salt used as a buffering agent, pH adjuster, and phosphate source in laboratory and pharmaceutical applications. It typically appears as a white crystalline powder with high purity and solubility in water, making it suitable for biochemical assays, media preparation, and formulation work requiring stable alkaline buffer capacity.
  • High purity (99.95%) suitable for analytical and formulation use.
  • Functions as a buffering agent and pH adjuster in aqueous systems.
  • Provides a soluble source of phosphate for media and reagent preparation.
  • Available in lab-scale pack sizes up to 500 g for routine use.
  • Stable, white crystalline powder with good water solubility.