Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Dipotassium hydrogen phosphate trihydrate | 16788-57-1 | MFCD00149926 | 99.2% | 228.22 g/mol | H7K2O7P | 1 KG
SDP

Catalog No. 5000208385
Encompass_Preferred

Dipotassium hydrogen phosphate trihydrate is an inorganic buffering salt used as a source of potassium and phosphate for preparing buffer solutions, cell culture media, and analytical reagents. It is supplied as a white to off-white solid with high purity and good water solubility.

  • High purity (99.2%).
  • Chemical formula H7K2O7P.
  • Molecular weight 228.22 g/mol.
  • White to off-white solid physical form.
  • Soluble in water (~50 mg/mL; may require ultrasonic assistance).
  • Available in 1 KG pack size.

Catalog No. 50-002-08385 Supplier Medchemexpress LLC Supplier No. HYW0880691KG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Dipotassium hydrogen phosphate trihydrate is an inorganic buffering salt used as a source of potassium and phosphate for preparing buffer solutions, cell culture media, and analytical reagents. It is supplied as a white to off-white solid with high purity and good water solubility.

  • High purity (99.2%).
  • Chemical formula H7K2O7P.
  • Molecular weight 228.22 g/mol.
  • White to off-white solid physical form.
  • Soluble in water (~50 mg/mL; may require ultrasonic assistance).
  • Available in 1 KG pack size.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.