Learn More
Medchemexpress LLC Dipotassium hydrogen phosphate trihydrate | 16788-57-1 | MFCD00149926 | 99.2% | 228.22 g/mol | H7K2O7P | 100 G
SDP
Catalog No. 5000208438
Encompass_Preferred
Dipotassium hydrogen phosphate trihydrate is an inorganic salt used as a buffering agent and a source of potassium and phosphate for laboratory and analytical applications. It is supplied as a solid trihydrate and commonly used in buffer preparation, media formulation, and biochemical assays.
  • Solid trihydrate form for stable handling and storage.
  • High purity suitable for analytical and laboratory use.
  • Commonly used as a buffering agent and source of potassium and phosphate.
  • Available in multiple pack sizes, including 100 G.
Catalog No. 50-002-08438 Supplier Medchemexpress LLC Supplier No. HYW088069100G
Dipotassium hydrogen phosphate trihydrate is an inorganic salt used as a buffering agent and a source of potassium and phosphate for laboratory and analytical applications. It is supplied as a solid trihydrate and commonly used in buffer preparation, media formulation, and biochemical assays.
  • Solid trihydrate form for stable handling and storage.
  • High purity suitable for analytical and laboratory use.
  • Commonly used as a buffering agent and source of potassium and phosphate.
  • Available in multiple pack sizes, including 100 G.