Learn More
Medchemexpress LLC Dipotassium tetrachloroplatinate | 10025-99-7 | ≥99.0% | 415.09 | 1 G
SDP
Catalog No. 5000439022
Encompass_Preferred
Dipotassium tetrachloroplatinate is an important reagent for the preparation of other platinum coordination complexes. It can be used as a radiosensitizer to enhance the killing effect of hyperthermia, serves as a catalyst, and exhibits antitumor activity. For research use only.
  • Enhances the killing effect of hyperthermia.
  • Serves as a catalyst.
  • Exhibits antitumor activity.
  • For research use only.
Catalog No. 50-004-39022 Supplier Medchemexpress LLC Supplier No. HY1120301G
Dipotassium tetrachloroplatinate is an important reagent for the preparation of other platinum coordination complexes. It can be used as a radiosensitizer to enhance the killing effect of hyperthermia, serves as a catalyst, and exhibits antitumor activity. For research use only.
  • Enhances the killing effect of hyperthermia.
  • Serves as a catalyst.
  • Exhibits antitumor activity.
  • For research use only.