Learn More
Weber Scientific DIPPER SAMPLER 100ML 50CS
SDP
Catalog No. NC3343072
Encompass
Dynalon Sterile Dipper Sampler 100mL , Color, Clear, handle length 385mm
Catalog No. NC3343072 Supplier Weber Scientific Supplier No. 303806
Dynalon Sterile Dipper Sampler 100mL , Color, Clear, handle length 385mm