Learn More
BURKLE INC Dipping bottle, 33.81 oz. (1000 ml), hxØ 13.0 x 5.83 in. (330 x 148 mm), 7.05 lbs (3.2 kg)
Catalog No. 5000025868
Encompass_Preferred
Dipping bottle with glass container for sampling in sewage plants, for mud and water samples. For testing crude oil, liquid mineral oil products, fuels, special and test fuels, fuel oils, petroleum and liquid lubricants of Hazard Categories IIA and IIB. Completely made from non-spark producing materials. Dipping frame made of nickel-plated brass, easy-click closure made of electrically conductive polyamide.
Catalog No. 50-000-25868 Supplier BURKLE INC Supplier No. 53654000
Dipping bottle with glass container for sampling in sewage plants, for mud and water samples. For testing crude oil, liquid mineral oil products, fuels, special and test fuels, fuel oils, petroleum and liquid lubricants of Hazard Categories IIA and IIB. Completely made from non-spark producing materials. Dipping frame made of nickel-plated brass, easy-click closure made of electrically conductive polyamide.