Learn More
BURKLE INC Dipping bottle EX, MS, 33.81 oz. (1000 ml), Ø 3.50 in. (89 mm), 7.28 pd. (3,3 kg)
Catalog No. 5000025813
Encompass_Preferred
Dipping bottle for testing mineral oil products Category A, Explosion Categories IIA, IIB and IIC, crude oil, liquid mineral oil products, fuels, special and test fuels, fuel oils, petroleum and liquid lubricants. Completely made from non-spark producing materials, nickel-plated
Catalog No. 50-000-25813 Supplier BURKLE INC Supplier No. 53653000
Dipping bottle for testing mineral oil products Category A, Explosion Categories IIA, IIB and IIC, crude oil, liquid mineral oil products, fuels, special and test fuels, fuel oils, petroleum and liquid lubricants. Completely made from non-spark producing materials, nickel-plated