Learn More
BURKLE INC Dipping vessel, V2A, 33.81 oz. (1000 ml), hxØ 16.81x3.23 in. (427x82mm), 70.55 oz. (2 kg)
Catalog No. 5000025698
Encompass_Preferred
Dipping vessel for taking all-layer samples from tanks, tank cars, water courses, water purification plants.
Catalog No. 50-000-25698 Supplier BURKLE INC Supplier No. 53652000
Dipping vessel for taking all-layer samples from tanks, tank cars, water courses, water purification plants.