Learn More
Jorgensen Labs JorVet Dip Quick Stain, Fixative Only, 500 mL
SDP
Catalog No. NC1758257
Encompass_Preferred
JorVet Dip Quick Stain is excellent for use in blood cell differential counts or as a general cytology stain, and gives coloration similar to the standard Wright-Giemsa stain. The rapid 3-step procedure can be done right in the wide-mouth 180ml. container in 15 seconds.
This item is not returnable. View return policy
Catalog No. NC1758257 Supplier Jorgensen Labs Supplier No. J0322A1
JorVet Dip Quick Stain is excellent for use in blood cell differential counts or as a general cytology stain, and gives coloration similar to the standard Wright-Giemsa stain. The rapid 3-step procedure can be done right in the wide-mouth 180ml. container in 15 seconds.