Learn More
TARGETMOL CHEMICALS INC DIPQUO HYDROCHLORIDE 5MG
SDP
Catalog No. 502581585
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. DIPQUO hydrochloride is an activator of the bone marker alkaline phosphatase (ALP) with an EC50 of 6.27 uM in C2C12 cells. DIPQUO hydrochloride promotes mouse and human osteoblast differentiation via activation of p38 MAPK-(beta).
Catalog No. 50-258-1585 Supplier TARGETMOL CHEMICALS INC Supplier No. T11048L5MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. DIPQUO hydrochloride is an activator of the bone marker alkaline phosphatase (ALP) with an EC50 of 6.27 uM in C2C12 cells. DIPQUO hydrochloride promotes mouse and human osteoblast differentiation via activation of p38 MAPK-(beta).