Learn More
Abcam 3,3'-Dipropylthiacarbocyanine iodide, Fluorescent membrane dye, 250MG
Catalog No. 50996249
Encompass_Preferred
MW 520.49 g/mol. Carbocyanine dye sensitive to membrane potential. Fluorescence response to depolarization depends on the staining concentration and detection method.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-996-249 Supplier Abcam Supplier No. AB145270250MG
MW 520.49 g/mol. Carbocyanine dye sensitive to membrane potential. Fluorescence response to depolarization depends on the staining concentration and detection method.
The product is subject to the following: Abcam Restricted Use Statement