Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dipropyl Adipate 98.0+%, TCI America™
SDP

Catalog No. A093125ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A093125ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A093125ML Supplier TCI America Supplier No. A093125ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 106-19-4
Molecular Formula C12H22O4
Molecular Weight (g/mol) 230.304
MDL Number MFCD00048696
InChI Key NKOUWLLFHNBUDW-UHFFFAOYSA-N
Synonym Adipic Acid Dipropyl Ester
PubChem CID 7790
IUPAC Name dipropyl hexanedioate
SMILES CCCOC(=O)CCCCC(=O)OCCC

Specifications

Color Colorless
Boiling Point 155°C
Quantity 25 mL
Formula Weight 230.30
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Dipropyl Adipate
EINECSNumber (2)-0879
RTECSNumber AV1740000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.