Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Dipropyl disulfide | 629-19-6 | MFCD00009378 | 99.3% | 150.31 g/mol | C6H14S2 | 1 G
SDP

Catalog No. 5000280779
Encompass_Preferred

Dipropyl disulfide is an organosulfur compound supplied as a high-purity research standard for biochemical and metabolic studies, analytical reference work, and assay development. It is offered in liquid and ready-to-use solution formats for convenient laboratory handling.

  • High purity (99.3%).
  • Molecular formula C6H14S2; molecular weight 150.31 g/mol.
  • Available as 1 G liquid and larger pack sizes; also as 10 mM 1 mL in DMSO solutions.
  • Storage: pure form -20°C (up to 3 years); in solvent -80°C (6 months).
  • Suitable for metabolic studies and analytical reference use.

Catalog No. 50-002-80779 Supplier Medchemexpress LLC Supplier No. HYW1968031G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Dipropyl disulfide is an organosulfur compound supplied as a high-purity research standard for biochemical and metabolic studies, analytical reference work, and assay development. It is offered in liquid and ready-to-use solution formats for convenient laboratory handling.

  • High purity (99.3%).
  • Molecular formula C6H14S2; molecular weight 150.31 g/mol.
  • Available as 1 G liquid and larger pack sizes; also as 10 mM 1 mL in DMSO solutions.
  • Storage: pure form -20°C (up to 3 years); in solvent -80°C (6 months).
  • Suitable for metabolic studies and analytical reference use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.