Learn More
Medchemexpress LLC Dipropyl disulfide | 629-19-6 | MFCD00009378 | 99.3% | 150.31 g/mol | C6H14S2 | 1 ML
SDP
Catalog No. 5000283198
Encompass_Preferred
Dipropyl disulfide is supplied as a ready-to-use 10 mM solution in DMSO (1 mL) for research applications. The compound has CAS number 629-19-6 and a reported purity of 99.3%. It is metabolized by flavin-containing monooxygenases and cytochrome P450 enzymes to produce metabolites such as propylglutathione sulfide conjugate and propylthiol. Intended for laboratory research use only.
  • Ready-to-use 10 mM solution in DMSO.
  • High reported purity (99.3%).
  • Useful for metabolic and enzymatic studies.
  • Includes CAS identifier for unambiguous identification.
  • For research use only; not for clinical or human use.
Catalog No. 50-002-83198 Supplier Medchemexpress LLC Supplier No. HYW19680310MM/1ML
Dipropyl disulfide is supplied as a ready-to-use 10 mM solution in DMSO (1 mL) for research applications. The compound has CAS number 629-19-6 and a reported purity of 99.3%. It is metabolized by flavin-containing monooxygenases and cytochrome P450 enzymes to produce metabolites such as propylglutathione sulfide conjugate and propylthiol. Intended for laboratory research use only.
  • Ready-to-use 10 mM solution in DMSO.
  • High reported purity (99.3%).
  • Useful for metabolic and enzymatic studies.
  • Includes CAS identifier for unambiguous identification.
  • For research use only; not for clinical or human use.