Learn More
Medchemexpress LLC Dipropyl disulfide | 629-19-6 | MFCD00009378 | 99.3% | 150.31 g/mol | C6H14S2 | 5 G
SDP
Catalog No. 5000280780
Encompass_Preferred
Dipropyl disulfide (CAS 629-19-6) is an organic dialkyl disulfide used as a research reagent and analytical standard. It is metabolized to dipropyl thiosulfinate by microsomal enzymes and is used in metabolism and analytical studies. Certificate of analysis and safety data sheet are provided for quality and safety documentation.
  • Chemical formula C6H14S2 and molecular weight 150.31 g/mol.
  • High purity of 99.3%.
  • Available in multiple pack sizes, including 5 g.
  • COA and SDS documentation available for each lot.
  • Suitable for research and analytical applications.
Catalog No. 50-002-80780 Supplier Medchemexpress LLC Supplier No. HYW1968035G
Dipropyl disulfide (CAS 629-19-6) is an organic dialkyl disulfide used as a research reagent and analytical standard. It is metabolized to dipropyl thiosulfinate by microsomal enzymes and is used in metabolism and analytical studies. Certificate of analysis and safety data sheet are provided for quality and safety documentation.
  • Chemical formula C6H14S2 and molecular weight 150.31 g/mol.
  • High purity of 99.3%.
  • Available in multiple pack sizes, including 5 g.
  • COA and SDS documentation available for each lot.
  • Suitable for research and analytical applications.