Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dipropyl Malonate 98.0+%, TCI America™
SDP

Catalog No. M003425G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M003425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M003425G Supplier TCI America Supplier No. M003425G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 1117-19-7
Molecular Formula C9H16O4
Molecular Weight (g/mol) 188.223
MDL Number MFCD00059406
InChI Key LWIWFCDNJNZEKB-UHFFFAOYSA-N
Synonym Malonic Acid Dipropyl Ester
PubChem CID 517959
IUPAC Name dipropyl propanedioate
SMILES CCCOC(=O)CC(=O)OCCC

Specifications

Melting Point -77°C
Color Colorless
Boiling Point 229°C
Quantity 25 g
Formula Weight 188.22
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Dipropyl Malonate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.