Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dipropyl Sulfate 90.0+%, TCI America™
SDP

Catalog No. D1638500G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D1638500G 500 g
D163825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D1638500G Supplier TCI America Supplier No. D1638500G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 598-05-0
Molecular Formula C6H14O4S
Molecular Weight (g/mol) 182.234
MDL Number MFCD00059243
InChI Key JYCKNDWZDXGNBW-UHFFFAOYSA-N
PubChem CID 69015
IUPAC Name dipropyl sulfate
SMILES CCCOS(=O)(=O)OCCC

Specifications

Color Yellow
Boiling Point 115°C
Quantity 500 g
UN Number 2922
Formula Weight 182.23
Percent Purity ≥90.0% (GC)
Physical Form Liquid
Chemical Name or Material Dipropyl Sulfate [Alkylating Agent]
EINECSNumber (2)-1673
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.