Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dipropyl Sulfite 99.0+%, TCI America™
SDP

Catalog No. D093525ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D093525ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D093525ML Supplier TCI America Supplier No. D093525ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 623-98-3
Molecular Formula C6H14O3S
Molecular Weight (g/mol) 166.235
MDL Number MFCD00059404
InChI Key MAIQPVFXODAAIG-UHFFFAOYSA-N
Synonym di-n-propyl sulfite, di-n-propyl sulphite, sulfurous acid, dipropyl ester, dipropyl sulphite, dipropylsulfite, di-n-propylsulfit, sulfurous acid dipropyl ester, sulphurous acid dipropyl ester, dipropyl ester of sulfurous acid
PubChem CID 136434
IUPAC Name dipropyl sulfite
SMILES CCCOS(=O)OCCC

Specifications

Color Yellow
Boiling Point 192°C
Quantity 25 mL
Formula Weight 166.24
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material Dipropyl Sulfite
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.