Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dipropylamine Hydrochloride 99.0+%, TCI America™
SDP

Catalog No. D093125G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D093125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D093125G Supplier TCI America Supplier No. D093125G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 5326-84-1
Molecular Formula C6H16ClN
Molecular Weight (g/mol) 137.651
MDL Number MFCD00054288
InChI Key GAZIBGHLWYHBDT-UHFFFAOYSA-N
PubChem CID 12202988
IUPAC Name N-propylpropan-1-amine;hydrochloride
SMILES CCCNCCC.Cl

Specifications

Color White
Quantity 25 g
Formula Weight 137.65
Percent Purity ≥99.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Dipropylamine Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.