Learn More
Enzo Life Sciences Diprotin A (5 mg) CAS: 90614-48-5
SDP
Supplier:  Enzo Life Sciences ALX260036M005
Encompass_Preferred
DPP4 inhibitor. Inhibitor of dipeptidyl aminopeptidase IV. Alternative Name: H-Ile-Pro-Ile-OH. Formula: C17H31N3O4. MW: 341.5. CAS: 90614-48-5. Purity: ≥98% (HPLC). Appearance: White to off-white solid. Solubility: Soluble in water (1mg/ml). Long Term Storage: -20°C
Catalog No. 50-103-1733