Learn More
Medchemexpress LLC HY-111174A 10mg Medchemexpress, Diprotin A (TFA) CAS:209248-71-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY111174A10MG
Encompass_Preferred
Medchemexpress, HY-111174A 10mg Diprotin A (TFA) CAS:209248-71-5 Diprotin A (TFA) is an inhibitor of dipeptidyl peptidase IV (DPP-IV). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8828