Learn More
Cayman Chemical DIprovocIm-1 10mg
SDP
Catalog No. 502949045
Encompass_Preferred
An agonist of TLR1/TLR2; induces TNF-α release in THP-1 cells (EC50 = 110 pM); increases the production of ovalbumin-specific IgG1 in wild-type mice sensitized to ovalbumin at 10 mg/kg; enhances anti-PD-L1 antibody-induced activation of cytotoxic T lymphocytes, reduction of tumor growth, and increases in survival in a B16/F10 murine melanoma model
Catalog No. 50-294-9045 Supplier Cayman Chemical Supplier No. 3317410 MG
An agonist of TLR1/TLR2; induces TNF-α release in THP-1 cells (EC50 = 110 pM); increases the production of ovalbumin-specific IgG1 in wild-type mice sensitized to ovalbumin at 10 mg/kg; enhances anti-PD-L1 antibody-induced activation of cytotoxic T lymphocytes, reduction of tumor growth, and increases in survival in a B16/F10 murine melanoma model