Learn More
Cayman Chemical DIpsacosIde B 10mg
SDP
Catalog No. 502909973
Encompass_Preferred
A triterpenoid saponin with hepatoprotective activity; enhances acetaminophen-induced autophagy and inhibits acetaminophen-induced apoptosis in HepG2 cells at 0.5 µM; increases survival in a mouse model of acetaminophen-induced acute liver failure at 50 mg/kg
Catalog No. 50-290-9973 Supplier Cayman Chemical Supplier No. 3562410 MG
A triterpenoid saponin with hepatoprotective activity; enhances acetaminophen-induced autophagy and inhibits acetaminophen-induced apoptosis in HepG2 cells at 0.5 µM; increases survival in a mouse model of acetaminophen-induced acute liver failure at 50 mg/kg