Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Dipsacus saponin A | 99624-66-5 | 796.98 | 1 MG
SDP

Catalog No. 5000416429
Encompass_Preferred

Dipsacus saponin A is an orally active triterpenoid saponin found in the roots of Dipsacus asper. It promotes bone repair by regulating osteoblast activity and inhibiting osteoclast function. It is promising for research of bone diseases such as fracture healing and osteoporosis.

  • Purity: 99.38%
  • Appearance: solid
  • Color: white to off-white
  • Initial source: plants Dipsacaceae Dipsacus asper
  • Sold for research purposes only, not for human use or sale to patients.

Catalog No. 50-004-16429 Supplier Medchemexpress LLC Supplier No. HYN157231MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Dipsacus saponin A is an orally active triterpenoid saponin found in the roots of Dipsacus asper. It promotes bone repair by regulating osteoblast activity and inhibiting osteoclast function. It is promising for research of bone diseases such as fracture healing and osteoporosis.

  • Purity: 99.38%
  • Appearance: solid
  • Color: white to off-white
  • Initial source: plants Dipsacaceae Dipsacus asper
  • Sold for research purposes only, not for human use or sale to patients.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.