Learn More
Medchemexpress LLC Dipsanoside A | 889678-62-0 | 1 MG
SDP
Catalog No. 5000421658
Encompass_Preferred
Dipsanoside A is a novel tetrairidoid glucoside originating from Dipsacus asper. This perennial plant, prevalent in China, has a long history of use in traditional Chinese medicine.
  • Used as a tonic for refreshment
  • Acts as a fissiparism promoter of osseous cells
  • Functions as an embryo security agent
Catalog No. 50-004-21658 Supplier Medchemexpress LLC Supplier No. HYN22381MG
Dipsanoside A is a novel tetrairidoid glucoside originating from Dipsacus asper. This perennial plant, prevalent in China, has a long history of use in traditional Chinese medicine.
  • Used as a tonic for refreshment
  • Acts as a fissiparism promoter of osseous cells
  • Functions as an embryo security agent