Learn More
Kenlor Industries Inc Kenlor Dipper Urine Control Level 1 & Level 2 - 5 X 15 mL each
Catalog No. NC0495786
Encompass
Kenlor semi-quantitative dipstick liquid urine control has values for Leukocytes, Urobilinogen, Bilirubin, Glucose, Blood, pH, Nitrite, Ketones, Protein, & Specific Gravity, plus HCG, Creatine, Microalbumin. Stable for 2 years at 2-8°C, 30 days at room temperature. Stable for 30 days at room temperature or 120 days at 2-8°C after opening, or 30 uses.
Catalog No. NC0495786 Supplier Kenlor Industries Inc Supplier No. 260306
Kenlor semi-quantitative dipstick liquid urine control has values for Leukocytes, Urobilinogen, Bilirubin, Glucose, Blood, pH, Nitrite, Ketones, Protein, & Specific Gravity, plus HCG, Creatine, Microalbumin. Stable for 2 years at 2-8°C, 30 days at room temperature. Stable for 30 days at room temperature or 120 days at 2-8°C after opening, or 30 uses.