Learn More
Cayman Chemical DIpyrIdamole-d16 5mg
SDP
Catalog No. 502990489
Encompass_Preferred
An internal standard for the quantification of dipyridamole by GC- or LC-MS
Catalog No. 50-299-0489 Supplier Cayman Chemical Supplier No. 301385 MG
An internal standard for the quantification of dipyridamole by GC- or LC-MS