Learn More
Selleck Chemical LLC Diquafosol Tetrasodium
SDP
Catalog No. 5000002912
Encompass_Preferred
Diquafosol Tetrasodium (INS-365) a P2Y2 receptor agonist is a pharmaceutical drug for the treatment of dry eye disease
Catalog No. 50-000-02912 Supplier Selleck Chemical LLC Supplier No. S65925MG
Diquafosol Tetrasodium (INS-365) a P2Y2 receptor agonist is a pharmaceutical drug for the treatment of dry eye disease