Learn More
Cayman Chemical Diquafosol (sodium salt), Purity: ≥95%, CAS: 211427-08-6, 50 mg
SDP
Catalog No. 502950773
Encompass_Preferred
A P2Y2 and P2Y4 receptor agonist (EC50s = 0.1 and 0.4 µM, respectively, in calcium mobilization assays); selective for P2Y2 and P2Y4 receptors over P2Y6 receptors (EC50 = 20 µM); induces the release of mucin-like glycoproteins from rabbit conjunctival goblet cells, as well as reduces desiccation-induced corneal damage in a rabbit model of dry eye disease at 0.1 and 1% w/v
Catalog No. 50-295-0773 Supplier Cayman Chemical Supplier No. 3397150 MG
A P2Y2 and P2Y4 receptor agonist (EC50s = 0.1 and 0.4 µM, respectively, in calcium mobilization assays); selective for P2Y2 and P2Y4 receptors over P2Y6 receptors (EC50 = 20 µM); induces the release of mucin-like glycoproteins from rabbit conjunctival goblet cells, as well as reduces desiccation-induced corneal damage in a rabbit model of dry eye disease at 0.1 and 1% w/v