Learn More
Cayman Chemical Diquat-d12 (bromide), Purity: ≥99% deuterated forms (d1-d12), 10 mg
SDP
Catalog No. 502932668
Encompass_Preferred
An internal standard for the quantification of diquat by GC- or LC-MS
Catalog No. 50-293-2668 Supplier Cayman Chemical Supplier No. 2677810 MG
An internal standard for the quantification of diquat by GC- or LC-MS