Learn More
Cayman Chemical Diquat-d4 (bromide), Purity: ≥99% deuterated forms (d1-d4), 10 mg
SDP
Catalog No. 502979742
Encompass_Preferred
An internal standard for the quantification of diquat by GC- or LC-MS
Catalog No. 50-297-9742 Supplier Cayman Chemical Supplier No. 1825010 MG
An internal standard for the quantification of diquat by GC- or LC-MS