Learn More
Cayman Chemical DIquat-d4bromIde 5mg
SDP
Catalog No. 502980426
Encompass_Preferred
An internal standard for the quantification of diquat by GC- or LC-MS
Catalog No. 50-298-0426 Supplier Cayman Chemical Supplier No. 182505 MG
An internal standard for the quantification of diquat by GC- or LC-MS