Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Diquat dibromide | 85-00-7 | 99.9% | 1 ML
SDP

Catalog No. 5000233753
Encompass_Preferred

Diquat dibromide is a quaternary ammonium herbicide supplied as an analytical standard for research applications. This listing corresponds to a 10 mM solution supplied in a 1 mL vial (CAS 85-00-7). Molecular formula C12H12Br2N2; molecular weight 344.05 g/mol.

  • Analytical standard suitable for research and analytical use.
  • Provided as a 10 mM solution.
  • Packaged as a 1 mL vial.
  • CAS number 85-00-7 for unequivocal identification.
  • Molecular formula C12H12Br2N2 and molecular weight 344.05 g/mol.

Catalog No. 50-002-33753 Supplier Medchemexpress LLC Supplier No. HY12298410MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Diquat dibromide is a quaternary ammonium herbicide supplied as an analytical standard for research applications. This listing corresponds to a 10 mM solution supplied in a 1 mL vial (CAS 85-00-7). Molecular formula C12H12Br2N2; molecular weight 344.05 g/mol.

  • Analytical standard suitable for research and analytical use.
  • Provided as a 10 mM solution.
  • Packaged as a 1 mL vial.
  • CAS number 85-00-7 for unequivocal identification.
  • Molecular formula C12H12Br2N2 and molecular weight 344.05 g/mol.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.