Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Diquat dibromide | 85-00-7 | 99.9% | 100 MG
SDP

Catalog No. 5000233752
Encompass_Preferred

Diquat dibromide is the dibromide salt of diquat supplied as a research-grade analytical standard for laboratory use. It is a light yellow solid that promotes reactive oxygen species formation and is used in herbicide research. This material is for research use only and is not intended for human or patient use.

  • High purity (99.9%).
  • Molecular weight 344.05 g/mol.
  • Chemical formula C12H12Br2N2.
  • Appearance light yellow solid.
  • Storage sealed and away from moisture; in solvent: -80°C (6 months), -20°C (1 month).
  • Not for human or patient use; cytotoxic and neurotoxic.

Catalog No. 50-002-33752 Supplier Medchemexpress LLC Supplier No. HY122984100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Diquat dibromide is the dibromide salt of diquat supplied as a research-grade analytical standard for laboratory use. It is a light yellow solid that promotes reactive oxygen species formation and is used in herbicide research. This material is for research use only and is not intended for human or patient use.

  • High purity (99.9%).
  • Molecular weight 344.05 g/mol.
  • Chemical formula C12H12Br2N2.
  • Appearance light yellow solid.
  • Storage sealed and away from moisture; in solvent: -80°C (6 months), -20°C (1 month).
  • Not for human or patient use; cytotoxic and neurotoxic.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.