Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Diquat (dibromide) | 85-00-7 | 99.9% | 25 MG
SDP

Catalog No. 5000233754
Encompass_Preferred

Diquat dibromide is the dibromide salt of diquat, a non-selective contact herbicide supplied as a light yellow to yellow solid analytical standard for research and analytical applications.

  • Cas number 85-00-7
  • Molecular formula C12H12Br2N2
  • Molecular weight 344.05 g·mol⁻¹
  • High purity (99.9% by LCMS)
  • Supplied as a 25 MG solid
  • Storage: 4°C sealed, away from moisture; in solution -80°C (6 months) or -20°C (1 month)

Catalog No. 50-002-33754 Supplier Medchemexpress LLC Supplier No. HY12298425MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Diquat dibromide is the dibromide salt of diquat, a non-selective contact herbicide supplied as a light yellow to yellow solid analytical standard for research and analytical applications.

  • Cas number 85-00-7
  • Molecular formula C12H12Br2N2
  • Molecular weight 344.05 g·mol⁻¹
  • High purity (99.9% by LCMS)
  • Supplied as a 25 MG solid
  • Storage: 4°C sealed, away from moisture; in solution -80°C (6 months) or -20°C (1 month)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.