Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Diquat dibromide hydrate | 6385-62-2 | 99.92% | 362.06 | 100 MG
SDP

Catalog No. 5000349138
Encompass_Preferred

Diquat dibromide hydrate is a comprehensive herbicide. It increases the production of Reactive Oxygen Species (ROS) and triggers mitochondrial autophagy. It generates free radicals like superoxide anions through redox cycles, leading to oxidative stress. Diquat dibromide hydrate is cytotoxic, reproductive, and neurotoxic, and is used in crops such as cotton and soybean to control noxious weeds.

  • Appearance: Solid
  • Color: Light yellow to yellow
  • Formula: C12H14Br2N2O
  • Shipping: Room temperature in continental US; may vary elsewhere
  • Storage: 4°C, sealed storage, away from moisture. In solvent: -80°C for 6 months; -20°C for 1 month (sealed storage, away from moisture)
  • Solubility (In Vitro): 125 mg/mL (345.25 mM) in H2O (requires ultrasonic)

Catalog No. 50-003-49138 Supplier Medchemexpress LLC Supplier No. HY136372100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Diquat dibromide hydrate is a comprehensive herbicide. It increases the production of Reactive Oxygen Species (ROS) and triggers mitochondrial autophagy. It generates free radicals like superoxide anions through redox cycles, leading to oxidative stress. Diquat dibromide hydrate is cytotoxic, reproductive, and neurotoxic, and is used in crops such as cotton and soybean to control noxious weeds.

  • Appearance: Solid
  • Color: Light yellow to yellow
  • Formula: C12H14Br2N2O
  • Shipping: Room temperature in continental US; may vary elsewhere
  • Storage: 4°C, sealed storage, away from moisture. In solvent: -80°C for 6 months; -20°C for 1 month (sealed storage, away from moisture)
  • Solubility (In Vitro): 125 mg/mL (345.25 mM) in H2O (requires ultrasonic)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.